
Corning® Transwell® 12 well plates

Products
Product Category
Color
Manufacturer
Binding Type
Diameter (mm)
Surface Area cm²
Wells
Working Volume (ml)
Color
Membrane Material
Plate Material
Package Size
Pore size (µm)
Sterility
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing