78952
[(3R)-3-Hydroxytetradecanoyl]-L-carnitine-(N-methyl-d3)
analytical standard
Synonym(s):
(2R)-3-Carboxy-2-[[(3R)-3-hydroxy-1-oxotetradecyl]oxy]-N,N-dimethyl-N-(methyl-d3)-1-propanaminium inner salt
About This Item
Recommended Products
grade
analytical standard
Assay
≥95.0% (HPLC)
optical activity
[α]/D -27.0 to -21.0°, c = 0.1 in methanol
shelf life
limited shelf life, expiry date on the label
extent of labeling
≥99%
storage temp.
−20°C
SMILES string
C[N+](C)(C([2H])([2H])[2H])C[C@H](OC(C[C@@H](O)CCCCCCCCCCC)=O)CC([O-])=O
InChI
1S/C21H41NO5/c1-5-6-7-8-9-10-11-12-13-14-18(23)15-21(26)27-19(16-20(24)25)17-22(2,3)4/h18-19,23H,5-17H2,1-4H3/t18-,19+/m0/s1/i2D3
InChI key
GFAZJTUXMHGTOC-RGIFXTRNSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service