Skip to Content
Merck
All Photos(1)

Key Documents

75772

Supelco

Isoxaben

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24N2O4
CAS Number:
Molecular Weight:
332.39
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

2-8°C

SMILES string

CCC(C)(CC)c1cc(NC(=O)c2c(OC)cccc2OC)on1

InChI

1S/C18H24N2O4/c1-6-18(3,7-2)14-11-15(24-20-14)19-17(21)16-12(22-4)9-8-10-13(16)23-5/h8-11H,6-7H2,1-5H3,(H,19,21)

InChI key

PMHURSZHKKJGBM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate. Download your certificate at: http://www.sigma aldrich.com

Isoxaben is a pre-emergence broad-leaf herbicide used primarily on small grains, turf, and ornamentals. This systematic herbicide is absorbed primarily by roots and translocated to stems and leaves. Isoxaben is a highly selective and potent inhibitor of cellulose biosynthesis in plants. It inhibits the incorporation of glucose into the cellulose-rich acid-insoluble fraction of isolated cell walls and disrupts its growth.

Isoxaben is approved for use in European Union (EU) according to Regulation (EC) No 1107/2009 (repealing Directive 91/414/EEC). As per Reg. (EC) No 149/2008, maximum residue levels (MRLs) have been set for various products of plant and animal origin from 0.01 to 0.1 mg/kg.

Application

It is intended to be used as a certified reference material (CRM) for calibration in chromatography and other analytical techniques. Isoxaben CRM may also find its use as described below:
  • To measure the effect of isoxaben on the incorporation of radiolabeled glucose, leucine, uracil, and acetate into acid-insoluble cell wall material, protein, nucleic acids, and fatty acids, respectively, in Arabidopsis thaliana
  • To determine the relative toxicities of isoxaben and two of its primary metabolites in soybean and wheat cell cultures
  • To investigate the effect of trifluralin, pronamide, haloxyfop-p methyl, propaquizafop, and isoxaben on weed control and oilseed rape yield
  • To study the degradation of isoxaben in soil and an aqueous system followed by isolation and identification of degradation products formed
  • To examine the effect of four different organic wastes, composted sheep manure, spent coffee grounds, composted pine bark, and coir, on potential groundwater pollution of propanil, isoxaben, cadusafos, and pencycuron under laboratory conditions

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service