SRP5262
SETMAR, GST tagged human
recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
METNASE
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.32
Recommended Products
recombinant
expressed in baculovirus infected Sf9 cells
Assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
mol wt
66-69 kDa
NCBI accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... SETMAR(6419)
General description
SETMAR or SET domain and mariner transposase fusion gene is a nonhomologous end-joining repair protein that regulates genomic integration of exogenous DNA by opening chromatin and facilitating joining of DNA ends. SETMAR has histone methyltransferase activity and methylates ′Lys-4′ and ′Lys-36′ of histone H3. SETMAR also has DNA nicking activity and may play a role in DNA repair. Human Pso4 forms a stable complex with SETMAR that regulates Metnase function in DNA repair. SETMAR has sequence-specific DNA-binding activity and recognizes the 19-mer core of the 5′-terminal inverted repeats (TIRs) of the Hsmar1 element.
Physical form
Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.
Preparation Note
After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service