SRP3302
CARBOXYPEPTIDASE-B rat
recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)
Synonym(s):
Cpb1
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352204
NACRES:
NA.32
Recommended Products
biological source
rat
recombinant
expressed in E. coli
Assay
≥95% (HPLC)
≥95% (SDS-PAGE)
form
lyophilized
mol wt
35.1 kDa
packaging
pkg of 25 μg
impurities
<0.1 EU/μg endotoxin, tested
color
white to off-white
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
rat ... Cpb2(113936)
General description
Cpb2 (carboxypeptidase B2), also known as thrombin-activatable fibrinolysis, is secreted as a plasma zymogen known as procarboxypeptidase B. Recombinant rat Carboxypeptidase-B is a 35.1 kDa protein consisting of 307 amino acids.
Biochem/physiol Actions
Cpb2 (carboxypeptidase B2) enzyme catalyzes the release of basic amino acids, arginine and lysine, from the C-terminal of proteins and peptides. It also functions as an esterase against hippuryl-n-argininic acid.
Physical form
Lyophilized from 20 mM Tris, pH 8.0 + 50 mM NaCl.
Reconstitution
Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Carboxypeptidase B inhibitors reduce tissue factor-induced renal microthrombi in rats.
Muto Y, et al.
European Journal of Pharmacology, 461(2-3), 181-189 (2003)
The kinetics of carboxypeptidase B activity.
E C WOLFF et al.
The Journal of biological chemistry, 237, 3094-3099 (1962-10-01)
E Clauser et al.
The Journal of biological chemistry, 263(33), 17837-17845 (1988-11-25)
Nucleotide sequencing of a rat carboxypeptidase B (CPB) cDNA and direct sequencing of the CPB mRNA via primer extension on pancreatic polyadenylated RNA has yielded the complete amino acid sequence of rat CPB. The rat enzyme is synthesized as a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service