Key Documents
SAE0060
Sigma-Aldrich
OxyDish
About This Item
UNSPSC Code:
41121800
NACRES:
NA.54
material
polystyrene
Quality Level
sterility
sterile; γ-irradiated
feature
optically clear, flat bottom, for growing anaerobes
size
90 mm
Related Categories
Legal Information
OxyDish is a trademark of Oxyrase, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)