Skip to Content
Merck
All Photos(1)

Key Documents

G1513

Sigma-Aldrich

Glycerol formal

47-67% 5-hydroxy-1,3-dioxane basis (GC), 33-53% 4-hydroxymethyl-1,3-dioxolane basis (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O3
CAS Number:
Molecular Weight:
104.10
Beilstein:
103206
MDL number:
UNSPSC Code:
50161901
PubChem Substance ID:
NACRES:
NA.25

Quality Level

contains

~0.02% 2,6-di-tert-butyl-4-methylphenol as stabilizer

composition

5-hydroxy-1,3-dioxane, 47-67% GC
4-hydroxymethyl-1,3-dioxolane, 33-53% GC

refractive index

n20/D 1.451 (lit.)

bp

192-193 °C (lit.)

density

1.203 g/mL at 25 °C (lit.)

SMILES string

OCC1COCO1.OC2COCOC2

InChI

1S/2C4H8O3/c5-4-1-6-3-7-2-4;5-1-4-2-6-3-7-4/h2*4-5H,1-3H2

InChI key

JIUMSISXCKWZTA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glycerol formal (GlyF) includes isomers, namely, 4-hydroxymethyl-1,3-dioxolane and 5-hydroxy-1,3-dioxane. It is a glycerol synthon with a combination of two 5- and 6-membered ring isomers.

Application

Glycerol formal is used to to solubilize water-insoluble compounds for subsequent aqueous dilution. It has been used as a chemical and dye emulsifier and as a cosolvent for drug delivery. Glycerol formal was used as a vehicle for antibiotic delivery in rats.

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Selective catalytic etherification of glycerol formal and solketal with dialkyl carbonates and K 2 CO 3
Selva M, et al.
Green Chemistry, 188-200 (2012)
Properties of fatty acid glycerol formal ester (FAGE) for use as a component in blends for diesel engines
Lapuerta M, et al.
Biomass and Bioenergy, 130-140 (2015)
Sarah Nickolls et al.
Advances in pharmacological sciences, 2011, 608912-608912 (2011-12-14)
GABA(A) receptors containing α2/3 subunits are current targets in the battle to develop new pain medications, as they are expressed in the spinal cord where increasing inhibitory drive should result in analgesia. However, this approach is prone to a range
Kazuo Hotta et al.
Anticancer research, 35(9), 4681-4689 (2015-08-09)
We investigated whether hepatic multidrug resistance-associated protein 2 (ABCC2) is involved in the hepatobiliary excretion of regorafenib, a novel multi-kinase inhibitor, using Sprague-Dawley (SD) rats and Eisai hyperbilirubinemic rats (EHBR) lacking the efflux transporter ABCC2. The involvement of organic anion-transporting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service