Key Documents
EP5418707005
Eppendorf® Rotor FA-45-18-11
incl. lid, aerosol tight
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
feature
to hold 18 x 1.5/2.0 mL tubes
incl. lid, aerosol tight
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5418707005
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5418/5418R 
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)