CLS3836
Corning® TC treated 1536 well microplates
black, clear bottom, low base, w/o lid, sterile
Synonym(s):
1536 multiwell plates, 1536 well microplates, 1536 well microtiter plates, 1536 well plates
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.13
Recommended Products
material
clear wells
polystyrene plate
square wells
sterility
sterile
feature
lid: no
black, clear bottom
packaging
case of 100 ea
manufacturer/tradename
Corning 3836
size
1536 wells
well volume
12.5 μL
well working volume
5-8 μL
color
black
Looking for similar products? Visit Product Comparison Guide
Features and Benefits
- Superior performance compared to other available commercial microplates: lower CV values, excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
- Optimized for automation with bar coding and standard readers
- Surface treated for optimal cell attachment
- Flat bottom wells with lid are compatible with automated dispensers including the PerkinElmer® FlexDrop
- Sterile and certified nonpyrogenic
Legal Information
Corning is a registered trademark of Corning, Inc.
PerkinElmer is a registered trademark of PerkinElmer Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service