CLS354603
Corning® BioCoat™ Matrigel® Matrix Thin Layer Multiwell Plate
well size 6 wells, flat bottom clear wells, lid
Synonym(s):
Cell Culture Plate, plastic coated cell culture plate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122102
NACRES:
NB.13
material:
flat bottom clear wells
size:
6 well
sterility:
sterile
binding type:
Tissue Culture (TC)-treated surface
feature:
lid
Recommended Products
material
flat bottom clear wells
sterility
sterile
feature
lid
packaging
pkg of 5 ea (1 pack of 5)
manufacturer/tradename
Corning 354603
size
6 well
well size
6 wells
binding type
Tissue Culture (TC)-treated surface
Application
Cell culture
Legal Information
BioCoat is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Matrigel is a registered trademark of Corning, Inc.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service