Skip to Content
Merck
All Photos(1)

Key Documents

CESS

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

human blood

packaging

tube of 5 μg 92110206-DNA-5UG
pkg of vial of cells 92110206-1VL

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblast

products

Surface Immunoglobulin ( Ig), MHC Class II

receptors

Fc, complement

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human lymphoblast

Cell Line Description

Derived from a patient with myelomonocytic leukaemia and subsequently exposed to Epstein Barr Virus (EBV). This line is EBV positive and therefore presumably derived from a normal B lymphocyte. This cell line is also positive for surface Ig, Fc receptor, complement receptor, surface Ia and EBNA. Responds to T cell replacement factor by increasing the number of IgG secreting cells. The cells should be handled under laboratory containment level 2.

Application

T cell replacement factor (TRF) assays, B cell differentiation studies

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 10,11
D13S317: 12
D16S539: 12
D5S818: 11,12
D7S820: 10,12
THO1: 7,9.3
TPOX: 8,9
vWA: 16,17

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 2-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service