NIST129C
Low alloy steel
NIST® SRM® 129c, high sulfur (SAE 112)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
grade
certified reference material
Quality Level
packaging
pkg of 150 g
manufacturer/tradename
NIST®
format
matrix material
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Other Notes
Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Carbon (C), Manganese (Mn), Phosphorus (P), Sulfur (S), Silicon (Si), Copper (Cu), Nickel (Ni), Chromium (Cr), Vanadium (V), Molybdenum (Mo)
See certificate for values and more details at nist.gov/SRM.
Carbon (C), Manganese (Mn), Phosphorus (P), Sulfur (S), Silicon (Si), Copper (Cu), Nickel (Ni), Chromium (Cr), Vanadium (V), Molybdenum (Mo)
See certificate for values and more details at nist.gov/SRM.
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service