ERMBF438D
Maize GMO Standard VCO-1981-5
ERM®, certified reference material, 1% VCO-1981-5
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
certified reference material
Agency
ERM®
manufacturer/tradename
JRC
application(s)
pharmaceutical (small molecule)
format
matrix material
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
Analysis Note
For more information please see:
ERMBF438D
ERMBF438D
Legal Information
ERM is a registered trademark of European Commission
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service