55739
Gonyautoxin-2 and Gonyautoxin-3 solution
20 μg/g in acetic acid (nominal concentration), analytical standard
Synonym(s):
GTX 2&3, GTX-2 - GTX-3 mixture
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H17N7O8S
Molecular Weight:
395.35
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
analytical standard
Quality Level
shelf life
limited shelf life, expiry date on the label
concentration
20 μg/g in acetic acid (nominal concentration)
format
multi-component solution
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Paralytic Shellfish Toxin PST for the analysis of marine toxins in seafood.
Check out our complete range of marine toxins reference materials
Check out our complete range of marine toxins reference materials
Analysis Note
20 μg/g (total) in 5 mM acetic acid
the mixture is "not equilibrated".
the mixture is "not equilibrated".
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service