Skip to Content
Merck
All Photos(1)

Documents

34516

Supelco

Folpet-d4

PESTANAL®, analytical standard

Synonym(s):

2-(Trichloromethylsulfanyl)isoindole-d4-1,3-dione, 2-[(Trichloromethyl)thio]-1H-isoindole-1,3(2H)-dione-d4, N-(Trichloromethylthio)phthalimide-d4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9D4Cl3NO2S
CAS Number:
Molecular Weight:
300.58
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

SMILES string

ClC(Cl)(Cl)SN1C(C2=C([2H])C([2H])=C([2H])C([2H])=C2C1=O)=O

InChI

1S/C9H4Cl3NO2S/c10-9(11,12)16-13-7(14)5-3-1-2-4-6(5)8(13)15/h1-4H/i1D,2D,3D,4D

InChI key

HKIOYBQGHSTUDB-RHQRLBAQSA-N

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Acute 1 - Carc. 2 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiuhong Li et al.
PloS one, 14(10), e0223724-e0223724 (2019-10-11)
A definitive endodermal cell lineage is a prerequisite for the efficient generation of mature endoderm derivatives that give rise to organs, such as the pancreas and liver. We previously reported that the induction of mesenchymal definitive endoderm cells depends on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service