Skip to Content
Merck
All Photos(1)

Documents

8.41264

Sigma-Aldrich

Triisopropyl borate

for synthesis

Synonym(s):

Triisopropyl borate, Boric acid triisopropyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H21BO3
CAS Number:
Molecular Weight:
188.07
MDL number:
UNSPSC Code:
12352108
EC Index Number:
226-529-9
NACRES:
NA.22

vapor pressure

75 hPa ( 100 °C)

Quality Level

Assay

≥97.0% (GC)

form

liquid

bp

139-141 °C/1013 hPa

mp

-59 °C

transition temp

flash point 25.5 °C

density

0.82 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C9H21BO3/c1-7(2)11-10(12-8(3)4)13-9(5)6/h7-9H,1-6H3

InChI key

NHDIQVFFNDKAQU-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.817 - 0.821
Identity (IR): passes test

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

62.6 °F - closed cup

Flash Point(C)

17 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service