Skip to Content
Merck
All Photos(1)

Documents

8.40106

Sigma-Aldrich

N-Benzyl-N-methylaniline

for synthesis

Synonym(s):

N-Benzyl-N-methylaniline, Benzylmethylphenyl amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15N
CAS Number:
Molecular Weight:
197.28
UNSPSC Code:
12352116
EC Index Number:
210-375-4
NACRES:
NA.22

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

450 °C (DIN 51794)

potency

2130 mg/kg LD50, oral (Rat)

bp

311 °C/1013 hPa

mp

8 °C

transition temp

flash point 152 °C

solubility

0.009 g/L

density

1.046 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C14H15N/c1-15(14-10-6-3-7-11-14)12-13-8-4-2-5-9-13/h2-11H,12H2,1H3

InChI key

LXZGVFCKZRHKMU-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.042 - 1.046
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

305.6 °F

Flash Point(C)

152 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service