8.20236
Sodium butyrate
for synthesis
Synonym(s):
Sodium butyrate, Butyric acid sodium salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C4H7NaO2
CAS Number:
Molecular Weight:
110.09
MDL number:
UNSPSC Code:
12352107
EC Index Number:
205-857-6
NACRES:
NA.22
Recommended Products
Quality Level
form
powder
mp
244 °C
density
1.324 g/cm3 at 30 °C
storage temp.
2-30°C
SMILES string
[Na+].[O-]C(=O)CCC
InChI
1S/C4H8O2.Na/c1-2-3-4(5)6;/h2-3H2,1H3,(H,5,6);/q;+1/p-1
InChI key
MFBOGIVSZKQAPD-UHFFFAOYSA-M
Analysis Note
Assay (HClO₄): ≥ 98.0 %
Identity (IR): passes test
Identity (IR): passes test
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service