Skip to Content
Merck
All Photos(1)

Key Documents

557450

Sigma-Aldrich

Ruthenium Red

A blocker of transmembrane Ca2+ fluxes (IC₅₀ = 7 µM) that attenuates capsaicin-induced cation channel opening.

Synonym(s):

Ruthenium Red, Ammoniated Ruthenium Oxychloride, RR

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(NH3)5RuORu(NH3)4ORu(NH3)5]Cl6
CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

description

Merck USA index - 14, 8300

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

red-black

solubility

ammonia: soluble
water: soluble

shipped in

ambient

storage temp.

10-30°C

General description

A blocker of transmembrane Ca2+ fluxes (IC50 = 7 µM) that attenuates capsaicin-induced cation channel opening. Inhibits Ca2+-induced Ca2+ release from ryanodine-sensitive intracellular Ca2+ stores. Inhibits K+-stimulated catecholamine secretion (IC50 = 6 µM).

Biochem/physiol Actions

Cell permeable: no
Primary Target
Blocker of transmembrane Ca2+ fluxes
Product does not compete with ATP.
Reversible: no
Target IC50: 7 µM in blocking transmembrane Ca2+ fluxes; 6 µM inhibiting K+-stimulated catecholamine secretion

Warning

Toxicity: Harmful (C)

Other Notes

Merck Index 13, 8379.
Phillippe, M. and Basa, A. 1996. Biochem. Biophys. Res. Commun. 221, 656.
Buratti, R., et al. 1995. Biochem. Biophys. Res. Commun. 213, 1082.
Hamilton, M.G. and Lundy, P.M. 1995. J. Pharmacol. Exp. Ther. 273, 940.
Gomis, A., et al. 1994. Biochem. Pharmacol. 47, 225.
Stjame, P., et al. 1994. Acta Otolaryngol. 114, 651.
Ma, J. 1993. J. Gen. Physiol.102, 1031.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service