Skip to Content
Merck
All Photos(1)

Key Documents

479975

Sigma-Aldrich

NDGA

A cell-permeable antioxidant and a selective lipoxygenase (LOX) inhibitor (IC₅₀ = 200 nM, 30 µM, and 30 µM for 5-LOX, 12-LOX, and 15-LOX, respectively) over cyclooxygenase (IC₅₀ = 100 µM).

Synonym(s):

NDGA, Nordihydroguaiaretic Acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22O4
CAS Number:
Molecular Weight:
302.36
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

description

Merck USA index - 14, 6693

Assay

≥90% (HPLC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

DMSO: 10 mg/mL
ethanol: soluble

shipped in

ambient

storage temp.

10-30°C

SMILES string

Oc1c(ccc(c1)C[C@H]([C@H](Cc2cc(c(cc2)O)O)C)C)O

InChI

1S/C18H22O4/c1-11(7-13-3-5-15(19)17(21)9-13)12(2)8-14-4-6-16(20)18(22)10-14/h3-6,9-12,19-22H,7-8H2,1-2H3/t11-,12+

InChI key

HCZKYJDFEPMADG-TXEJJXNPSA-N

General description

A cell-permeable antioxidant and a selective lipoxygenase (LOX) inhibitor (IC50 = 200 nM, 30 µM, and 30 µM for 5-LOX, 12-LOX, and 15-LOX, respectively) over cyclooxygenase (IC50 = 100 µM). Also shown to inhibit platelet-derived growth factor (PDGF)-stimulated DNA synthesis in Swiss/3T3 cells. A powerful hydroxy radical quencher.
A cell-permeable, antioxidant and a selective lipoxygenase (LO) inhibitor (IC50 = 200 nM, 30 µM, and 30 µM for 5-LO, 12-LO, and 15-LO, respectively) over cyclooxygenase (IC50 = 100 µM). Also shown to inhibit platelet-derived growth factor (PDGF)-stimulated DNA synthesis in Swiss 3T3 cells.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
LOX-5
Product does not compete with ATP.
Reversible: no
Target IC50: 200 nM, 30 µM, and 30 µM for 5-LOX, 12-LOX, and 15-LOX, respectively

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Domin, J., et al. 1994. J. Biol. Chem. 269, 8260.
Goodman, Y., et al. 1994. Brain Res. 654, 171.
Kozubik, A., et al. 1993. Exp. Hematol. 21, 138.
Salari, H., et al. 1984. Prostagland. Leuk. Med. 13, 53.
Hope, W.C., et al. 1983. Biochem. Pharmacol. 32, 367.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service