Skip to Content
Merck
All Photos(1)

Key Documents

239825

Sigma-Aldrich

AMD3100

InSolution, ≥98%, 50 mM aqueous solution, CXCR4 antagonist I

Synonym(s):

InSolution CXCR4 Antagonist I, AMD3100, 1,1ʹ(1,4-Phenylenebis(methylene))-bis-1,4,8,11-tetraazacyclotetradecane, 8HCl, 4H₂O, 1,4-bis((1,4,8,11-Tetraazacyclotetradecan-1-yl)methyl)benzene, 8HCl, 4H₂O

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H54N8 · 8HCl · 4H2O
CAS Number:
Molecular Weight:
866.53
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
protect from light

shipped in

dry ice

storage temp.

−70°C

SMILES string

Cl.Cl.Cl.Cl.Cl.Cl.Cl.Cl.N3(CCNCCCNCCNCCC3)Cc1ccc(cc1)CN2CCNCCCNCCNCCC2

InChI

1S/C28H54N8.8ClH/c1-9-29-15-17-31-13-3-21-35(23-19-33-11-1)25-27-5-7-28(8-6-27)26-36-22-4-14-32-18-16-30-10-2-12-34-20-24-36;;;;;;;;/h5-8,29-34H,1-4,9-26H2;8*1H

InChI key

UEUPDYPUTTUXLJ-UHFFFAOYSA-N

General description

A symmetrical bicyclam compound that antagonizes CXCL12 (SDF1) binding to CXCR4 (IC50 = 108 and 245 nM using rat and human CXCR4, respectively) and inhibits SDF1-induced Ca2+ flux (by 100% at 100 ng/ml in SUP-T1 and THP-1 cultures) as well as CXCR4-, but not CCR5-, mediated HIV infection (IC50 = 200 nM), while enhancing the binding of SDF1 to CXCR7 (by ~60% at 1 mM with CXCR7-expressing HEK293T cells) and SDF1-induced β-arrestin recruitment to CXCR7 (EC50 = 6.48 and 11.8 nM, in the presence and absence of 10 µM AMD3100, respectively). Administration AMD3100 via intravenous infusion is also reported to result in hematopoietic stem cell mobilization in humans, dogs, and mice in vivo. Exhibits no inhibitory effects against chemokine-induced signaling via CXCR1/2/3 or CCR1/2/3/4/5/6/7/8/9.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
CXCR4
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

A 50 mM (15 mg/308 µL) solution of CXCR4 Antagonist I, AMD3100 (Cat. No. 239820) in H₂O.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C). Aliquots are stable for up to 1 year at -70°C.

Other Notes

Kalatskaya, I., et al. 2009. Mol. Pharmacol.77, 1240.
Pitchford, S.C., et al. 2009. Cell Stem Cell4, 62.
De Clercq, E. 2009. Biochem. Pharmacol.77, 1655.
Thoma, G., et al. 2008. J. Med. Chem.51, 7915.
De Clercq, E. 2000. Mol. Pharmacol.57, 833.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service