1.07998
1,8-Dihydroxy-2-(4-sulfophenylazo)naphthalene-3,6- disulfonic acid trisodium salt
GR for analysis (reagent for fluoride, zirconium, thorium)
Synonym(s):
1,8-Dihydroxy-2-(4-sulfophenylazo)naphthalene-3,6- disulfonic acid trisodium salt, SPADNS
About This Item
Quality Level
pH
6.0 (22 °C, 10 g/L in H2O)
solubility
220 g/L
bulk density
810 kg/m3
storage temp.
15-25°C
InChI
1S/C16H12N2O11S3.3Na/c19-12-7-11(31(24,25)26)5-8-6-13(32(27,28)29)15(16(20)14(8)12)18-17-9-1-3-10(4-2-9)30(21,22)23;;;/h1-7,17,19H,(H,21,22,23)(H,24,25,26)(H,27,28,29);;;/q;3*+1/p-3/b18-15-;;;
InChI key
LSAWBVPBZOEGOT-ZOVWSQMSSA-K
Looking for similar products? Visit Product Comparison Guide
Related Categories
Analysis Note
Color change with zirconium: passes testpink - violet
Absorption maximum λmax. (buffer pH 7.0): 505 - 510 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.02 g/l; buffer pH 7.0; calc. on dried substance): 400 - 560
Loss on Drying (105°C): ≤ 10 %
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service