Skip to Content
Merck
All Photos(2)

Key Documents

850397C

Avanti

20:4 (Cis) PC

1,2-diarachidonoyl-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1,2-di-(5Z,8Z,11Z,14Z-eicosatetraenoyl)-sn-glycero-3-phosphocholine; DAPC; PC(20:4(5Z,8Z,11Z,14Z)/20:4(5Z,8Z,11Z,14Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H80NO8P
CAS Number:
Molecular Weight:
830.12
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850397C-25mg)
pkg of 2 × 4 mL (850397C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850397C

concentration

10 mg/mL (850397C-25mg)
25 mg/mL (850397C-200mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[P](=O)([O-])(OC[C@H](OC(=O)CCC\C=C/C\C=C/C\C=C/C\C=C\CCCCC)COC(=O)CCC\C=C/C\C=C/C\C=C/C\C=C/CCCCC)OCC[N+](C)(C)C

InChI

1S/C48H80NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-38-40-47(50)54-44-46(45-56-58(52,53)55-43-42-49(3,4)5)57-48(51)41-39-37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h14-17,20-23,26-29,32-35,46H,6-13,18-19,24-25,30-31,36-45H2,1-5H3/b16-14-,17-15+,22-20-,23-21-,28-26-,29-27-,34-32-,35-33-/t46-/m1/s1

InChI key

LZLVZIFMYXDKCN-HLKGGLNDSA-N

General description

20:4 (cis) PC or 1,2-diarachidonoyl-sn-glycero-3-phosphocholine is an unsaturated glycerophospholipid, with substitution of two chains of arachidonic acid at sn-1 and sn-2 positions of the glycerol backbone, along with choline as a headgroup.
The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.

Application

20:4 (cis) PC or 1,2-diarachidonoyl-sn-glycero-3-phosphocholine might be used:
  • in lipid bilayer, to study its effect on membrane vesiculation by dynamin and endophilin
  • as a phosphatidylcholine (PC) standard to compare the extraction efficiency of the three tested solvent systems prior to liquid chromatography electrospray ionization tandem mass spectrometry (LC-ESI-MS/MS) analysis
  • in low density lipid (LDL)-mimicking liposome as an alternative to LDL for evaluating the LDL-oxidation preventing antioxidants

Packaging

5 mL Clear Glass Sealed Ampule (850397C-200mg)
5 mL Clear Glass Sealed Ampule (850397C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seiji Takeda et al.
Biochemical and biophysical research communications, 513(1), 275-279 (2019-04-08)
Oxidation of low-density lipoproteins (LDL) causes atherosclerosis. Detection of oxidation of LDL-mimetic liposomes using an electrode might serve as a convenient tool in the search of antioxidants for the prevention of atherosclerosis. This report proposes a reaction mechanism between LDL-mimetic
Ana Gonzálvez et al.
Analytical and bioanalytical chemistry, 396(8), 2965-2975 (2010-02-27)
A zirconia (ZrO(2))-modified solid-phase extraction sorbent has been evaluated for selective extraction of phosphatidylcholines from biological samples, followed by analysis of the isolated solutes by reversed-phase liquid chromatography-electrospray ionization-tandem mass spectrometry. The clean-up process was optimized using seven standard phosphatidylcholines
Marco M Manni et al.
eLife, 7 (2018-03-16)
Phospholipid membranes form cellular barriers but need to be flexible enough to divide by fission. Phospholipids generally contain a saturated fatty acid (FA) at position sn1 whereas the sn2-FA is saturated, monounsaturated or polyunsaturated. Our understanding of the impact of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service