Skip to Content
Merck
All Photos(1)

Key Documents

810210P

Avanti

NBD 18:0 Ceramide

Avanti Research - A Croda Brand 810210P, powder

Synonym(s):

omega(7-nitro-2-1,3-benzoxadiazol-4-yl)-N-stearoyl-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H73N5O6
CAS Number:
Molecular Weight:
744.06
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 100 μg (with stopper and crimp cap (810210P-100ug))

manufacturer/tradename

Avanti Research - A Croda Brand 810210P

shipped in

dry ice

storage temp.

−20°C

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (810210P-100ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ieva Vasiliauskaité-Brooks et al.
Nature, 544(7648), 120-123 (2017-03-23)
Adiponectin receptors (ADIPORs) are integral membrane proteins that control glucose and lipid metabolism by mediating, at least in part, a cellular ceramidase activity that catalyses the hydrolysis of ceramide to produce sphingosine and a free fatty acid (FFA). The crystal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service