Z277703
HGS Polyhedron molecular model
student set, fundamental organic chemistry
All Photos(1)
About This Item
UNSPSC Code:
41100000
NACRES:
NB.17
Recommended Products
description
scale: 1 Å = 2.5 cm
feature
no. of atoms 52
no. of bonds 62
manufacturer/tradename
HGS 1000
Related Categories
General description
The use of 14, 20, and 26-sided polyhedrons as atomic nuclei makes these models unique. The various bond angles and distances needed for the models can be precisely represented, and the symmetrical characteristics of the structures can be observed.
- Color-coded polyhedrons to indicate different elements
- Flexible polyacetal connecting bonds permit easy assembly of complicated molecules and models of strained compounds and distorted crystals
- Å scale provided with sets allows measurement of bond distances
- Bond puller included for easy disassemble
- Two types of orbital plates for p atomic orbitals and π molecular orbitals (some sets only)
Choose from one of the most recent versions:
Certificates of Analysis (COA)