Z104485
Tubing quick-disconnects
for tubing i.d., 1/8 ‑ 1/4 in.
All Photos(1)
About This Item
Recommended Products
material
PE
feature
for tubing i.d. 1/8 ‑ 1/4 in.
packaging
pack of 12 ea
manufacturer/tradename
Bel-Art H19727-0000
General description
Consists of two serrated connectors which are joined securely through a male-female center taper. Connector of one size can be interchanged with other sizes to produce reducing combinations.
- Easy-to-assemble barbed connectors require no tools
- Ideal for use with flexible tubing
- Handy on lines that require frequent disconnection