DWKKC92812G-2000
KIMBLE® KIMAX® Heavy duty volumetric wide-mouth flask with KIMCOTE® safety coating and glass stopper
glass flask, capacity 2,000 mL, class A
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Recommended Products
material
glass flask
feature
class A
packaging
case of 1 ea
manufacturer/tradename
Kimble® KC92812G-2000
capacity
2,000 mL
General description
KimCote Class A Heavy Duty Wide Mouth Volumetric Flasks
- Pipet access is easy with wide-mouth volumetric flasks
- Superior strength, durability and safety from heavy, uniform walls
- Quick identification with large, permanent, easy-to-read markings
- Flat interior bottoms for stirring with standard stir bars
- Graduated to Class A volumetric tolerances
- Standard Taper glass stoppers
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Features and Benefits
• Complies with ASTM Specification E288, Class A requirements• Manufactured from borosilicate glass 3.3• Autoclavable KimCote safety coating reduces the hazards of shattered glass and leakage of toxic or corrosive chemicals• Wide mouth volumetric flasks feature neck dimensions that will easily accommodate pipets• Superior strength, durability and heavy uniform walls ensure safety in the lab• Calibrated to contain• Flat interior bottoms are ideal for stirring with standard stir bars
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
KIMCOTE is a registered trademark of DWK LIFE SCIENCES LLC
Kimax is a registered trademark of Gerresheimer Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)