ALD01648
3-methyl-3-((perfluorophenyl) amino)butan-1-ol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
form
oil
storage temp.
2-8°C
SMILES string
FC1=C(F)C(F)=C(NC(C)(C)CCO)C(F)=C1F
InChI
1S/C11H12F5NO/c1-11(2,3-4-18)17-10-8(15)6(13)5(12)7(14)9(10)16/h17-18H,3-4H2,1-2H3
InChI key
OONDYVAWDHEFLV-UHFFFAOYSA-N
Application
Traditionally challenging to access, this hindered amine provides a chemically differentiated building block for organic synthesis and medicinal chemistry, specifically for preparation of drug candidates containing hindered amine motifs. Additionally, this and over 80 other sterically hindered secondary amines in the Sigma-Aldrich portfolio provide structurally distinct screening compounds. Screen several through Aldrich Market Select. This hindered amine was accessed via a recent publication reported by Baran and coworkers using an innovative hydroamination method.
related product
Product No.
Description
Pricing
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service