936456
Diisopropanolethanolamine
≥95%
Synonym(s):
1,1′-((2-Hydroxyethyl)azanediyl)bis(propan-2-ol), 1,1′-[(2-Hydroxyethyl)imino]dipropan-2-ol, 1-[2-Hydroxyethyl(2-hydroxypropyl)amino]propan-2-ol, Di-(2-hydroxypropyl)-ethanolamine
About This Item
Quality Level
Assay
≥95%
form
viscous liquid
color
Colorless to Yellow
bp
152 °C
density
1.043 g/cm3
storage temp.
2-8°C
SMILES string
OCCN(CC(O)C)CC(O)C
InChI
1S/C8H19NO3/c1-7(11)5-9(3-4-10)6-8(2)12/h7-8,10-12H,3-6H2,1-2H3
InChI key
HHKUQCFQGCCLGA-UHFFFAOYSA-N
Application
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
354.2 °F
Flash Point(C)
179 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service