Skip to Content
Merck
All Photos(1)

Key Documents

903507

Sigma-Aldrich

Kevetrin-(4-isothioureido-13C,15N2-butyronitrile-13C,15N) hydrochloride

≥98 atom % 15N, ≥98 atom % 13C, ≥95% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C2C3H915N3SHCl
Molecular Weight:
184.64
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.24

isotopic purity

≥98 atom % 13C
≥98 atom % 15N

Quality Level

Assay

≥95% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

mass shift

M+5

storage temp.

−20°C

SMILES string

[15NH2][13C](SCCC[13C]#[15N])=[15NH].Cl

InChI key

NCXJZJFDQMKRKM-CBWHUSCUSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service