Skip to Content
Merck
All Photos(1)

Documents

719676

Sigma-Aldrich

2-Butoxyethanol-13C6

99 atom % 13C, 97% (CP)

Synonym(s):

Ethylene glycol butyl ether-13C6

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CH3(13CH2)3O13CH213CH2OH
CAS Number:
Molecular Weight:
124.13
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

97% (CP)

form

liquid

bp

169-172.5 °C (lit.)

density

0.947 g/mL at 25 °C

mass shift

M+6

SMILES string

[13CH3][13CH2][13CH2][13CH2]O[13CH2][13CH2]O

InChI

1S/C6H14O2/c1-2-3-5-8-6-4-7/h7H,2-6H2,1H3/i1+1,2+1,3+1,4+1,5+1,6+1

InChI key

POAOYUHQDCAZBD-IDEBNGHGSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

152.6 °F - closed cup

Flash Point(C)

67 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service