Skip to Content
Merck
All Photos(1)

Key Documents

680087

Sigma-Aldrich

(R)-(–)-2-(Trifluoromethyl)pyrrolidine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8F3N
CAS Number:
Molecular Weight:
139.12
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Pricing and availability is not currently available.

Quality Level

Assay

97%

form

solid

bp

110-111 °C

mp

29-33 °C

functional group

fluoro

storage temp.

2-8°C

SMILES string

FC(F)(F)[C@H]1CCCN1

InChI

1S/C5H8F3N/c6-5(7,8)4-2-1-3-9-4/h4,9H,1-3H2/t4-/m1/s1

InChI key

VINAMCOZNJHNIH-SCSAIBSYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
611387551384612456
format

single component solution

format

single component solution

format

single component solution

format

single component solution

grade

NMR reference standard, analytical standard

grade

NMR reference standard, analytical standard

grade

NMR reference standard, analytical standard

grade

NMR reference standard, analytical standard

concentration

0.1 M in DMSO-d6 (99.9 atom % D)

concentration

0.1 M in DMSO-d6 (99.9 atom % D)

concentration

90% in DMSO-d6 (99.9 atom % D)

concentration

0.1% in chloroform-d (99.8 atom % D)

NMR tube size

8 mm × 7 in.

NMR tube size

5 mm × 7 in.

NMR tube size

5 mm × 8 in.

NMR tube size

8 mm × 7 in.

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

84.0 °F - closed cup

Flash Point(C)

28.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Don't see the Right Version?

    If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service