Skip to Content
Merck
All Photos(1)

Key Documents

537276

Sigma-Aldrich

4-(Benzyloxy)benzyl chloride, polymer-bound

100-200 mesh, extent of labeling: 0.5-1.5 mmol/g loading, 1 % cross-linked with divinylbenzene

Synonym(s):

Benzyloxybenzyl chloride, polymer-bound, Wang chlorinated resin, [4-(Chloromethyl)phenoxymethyl]polystyrene cross-linked with divinylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352128
PubChem Substance ID:
NACRES:
NA.22

Quality Level

crosslinking

1 % cross-linked with divinylbenzene

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

extent of labeling

0.5-1.5 mmol/g loading

particle size

100-200 mesh

functional group

alcohol

storage temp.

2-8°C

SMILES string

C=Cc1ccccc1.C=Cc2ccc(C=C)cc2.ClCc3ccc(OCc4ccccc4)cc3

InChI key

YHHMDQVMLAVWTB-UHFFFAOYSA-N

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service