486957
Phenol-3,5-d2
97 atom % D
Synonym(s):
3,5-Dideuteriophenol
About This Item
Recommended Products
isotopic purity
97 atom % D
form
solid
bp
182 °C (lit.)
mp
40-42 °C (lit.)
density
1.094 g/mL at 25 °C
mass shift
M+2
SMILES string
[2H]c1cc([2H])cc(O)c1
InChI
1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H/i2D,3D
InChI key
ISWSIDIOOBJBQZ-PBNXXWCMSA-N
Packaging
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2
Target Organs
Nervous system,Kidney,Liver,Skin
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 2
Flash Point(F)
174.2 °F - closed cup
Flash Point(C)
79 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service