Merck
  • Alteration of Porcine Serum Albumin Levels in Pork Meat by Marination in Kiwi or Pineapple Juice and Subsequent Pan Broiling.
Alteration of Porcine Serum Albumin Levels in Pork Meat by Marination in Kiwi or Pineapple Juice and Subsequent Pan Broiling.
Korean journal for food science of animal resources (2014-01-01)
Sung-Sil Moon, Dongwook Kim, Il-Suk Kim, Jun-Sang Ham, Beom-Young Park, Aera Jang
ABSTRACT
This study was conducted to evaluate the changes in porcine serum albumin (PSA), a major allergen, which occur when raw pork ham is marinated with kiwi or pineapple juice, and/or when the ham is pan broiled at 300℃ for 4 min after marination. In this study, raw pork ham was soaked for 4 h or 8 h in marinades containing commercial marinating sauce only, commercial marinating sauce and 7% kiwi juice, or commercial marinating sauce and 7% pineapple juice. When the meat was marinated and then pan-broiled, pork ham meat protein was significantly denatured and hydrolyzed, and the level of PSA in the meat was significantly reduced. The PSA contents of pork broiled without marination, pork that had been marinated in commercial marinating sauce alone, pork that had been marinated in commercial marinating sauce with kiwi juice, and pork that had been marinated in commercial marinating sauce with pineapple juice, were 95.4, 43.3, 14.3, and 5.4 ng/mL, respectively (p<0.05). Marinating with pineapple juice was more effective than marinating with kiwi juice; and marination for 8 h was more effective than marinating for 4 h. These results indicate that the level of PSA in pork ham is effectively reduced, when the meat is first marinated in sauces that contain kiwi or pineapple extracts for 8 h, rather than 4 h, and then cooked. Further study is needed to determine whether marinated pork meat reduces allergenicity in vivo, as well.
Sigma-Aldrich
Albumin from porcine serum, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Albumin from porcine serum, lyophilized powder, ≥98% (agarose gel electrophoresis)