Merck
  • Negative results of carcinogenicity bioassay of methyl carbazate in rats: significance for the toxicological evaluation of carbadox.
Negative results of carcinogenicity bioassay of methyl carbazate in rats: significance for the toxicological evaluation of carbadox.
Toxicology (1981-01-01)
R Truhaut, R Ferrando, J M Faccini, A M Monro
ABSTRACT
Methyl carbazate, a metabolite of carbadox in the rat, was administered orally to rats for 2 years. The compound was mixed in the diet at concentrations corresponding to dose levels of 0, 2.5, 5 and 10 mg/kg body wt/day. There was no evidence of any treatment-related toxicity or, in particular, of carcinogenicity.
Sigma-Aldrich
Methyl hydrazinocarboxylate, 97%