Merck
  • Effects of Dietary Inclusion of Seaweed, Heat Stress and Genetic Strain on Performance, Plasma Biochemical and Hematological Parameters in Laying Hens.
Effects of Dietary Inclusion of Seaweed, Heat Stress and Genetic Strain on Performance, Plasma Biochemical and Hematological Parameters in Laying Hens.
Animals : an open access journal from MDPI (2020-09-10)
Shima Borzouie, Bruce M Rathgeber, Cassie M Stupart, Janice MacIsaac, Leslie A MacLaren
ABSTRACT
This study was planned to investigate the effects of seaweed supplementation, genetic strain, heat stress and their interactions on laying hen performances, blood chemistry and hematology. In a short-term trial, laying hens of the two genetic lines Lohman LSL-Lite (White) and Lohman Brown-Lite (Brown) were supplemented with Chondrus crispus (CC) at 3% for 21 days, while a control group was not. In a long-term trial, the same two strains were assigned to control (0%), 3% red seaweed Chondrus crispus (CC) or 0.5% brown seaweed Ascophyllum nodosum (AN)-supplemented diets for 41 weeks, concluding with a four-week control or heat-stress period. The White hens displayed higher egg production and a lower feed/egg ratio. The short-term inclusion of CC significantly reduced the feed intake, weight gain and feed/egg ratio. The long-term seaweed intake affected the plasma albumin and gamma-glutamyl transferase (GGT) (p < 0.05), and there were significant strain-heat stress interactions; heat stress in the Brown birds was associated with reduced protein, globulin and glucose and increased cholesterol and GGT levels and higher heterophil-to-lymphocyte (H/L) ratios (p < 0.05) in response to heat stress (p < 0.05). In conclusion, a long-term seaweed supplementation affected the plasma protein and enzyme profiles, yet had little effect on hen leukocyte counts and the overall performance.
Sigma-Aldrich
Sodium hydrogen sulfate, technical grade