Merck
  • Molecularly Imprinted Nanogels Capable of Porcine Serum Albumin Detection in Raw Meat Extract for Halal Food Control.
Molecularly Imprinted Nanogels Capable of Porcine Serum Albumin Detection in Raw Meat Extract for Halal Food Control.
Analytical chemistry (2020-04-14)
Chehasan Cheubong, Aoi Yoshida, Yuki Mizukawa, Natsuki Hayakawa, Minako Takai, Takahiro Morishita, Yukiya Kitayama, Hirobumi Sunayama, Toshifumi Takeuchi
ABSTRACT
Accurate, simple, and valuable analytical methods for detection of food contamination are rapidly expanding to evaluate the validity of food product quality because of ethnic considerations and food safety. Herein molecularly imprinted nanogels (MIP-NGs), capable of porcine serum albumin (PSA) recognition, were prepared as artificial molecular recognition elements. The MIP-NGs were immobilized on a quartz crystal microbalance (QCM) sensor for detection of pork contamination in real beef extract samples. The MIP-NGs-based QCM sensor showed high affinity and excellent selectivity toward PSA compared to reference serum albumins from five different animals. The high PSA specificity of MIP-NGs led to the detection of pork contamination with a detection limit of 1% (v/v) in real beef extract samples. We believe the artificial molecular recognition materials prepared by molecular imprinting are a promising candidate for halal food control.
Sigma-Aldrich
Albumin from porcine serum, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Albumin from porcine serum, lyophilized powder, ≥98% (agarose gel electrophoresis)