ERMBF442C
GHB811 Cotton GMO Standard
(nominal 0.1%), ERM®, certified reference material
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
certified reference material
Quality Level
Agency
ERM®
quality
(nominal 0.1%)
manufacturer/tradename
JRC
application(s)
food and beverages
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
Analysis Note
For more information please click on the below product number:
ERMBF442C
ERMBF442C
Legal Information
ERM is a registered trademark of European Commission
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service