Skip to Content
Merck
All Photos(1)

Key Documents

43308

Supelco

MOSH/MOAH Standard Mix 

certified reference material, TraceCERT®, 10 components in toluene, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

concentration

150-600 mg/L each component in toluene

application(s)

food and beverages

format

multi-component solution

storage temp.

−20°C

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Application

This certified reference material (CRM) mix is intended for the use of LC coupled with GC-FID analysis to determine mineral oil hydrocarbons (MOH) in food or as migrants from packaging material or from equipment used for food processing.

This mix contains suitable internal standard components for quantification of both mineral oil saturated hydrocarbons (MOSH) and mineral oil aromatic hydrocarbons (MOAH), as well as markers for the start and the end of the corresponding MOSH and MOAH fractions. More detailed information about the specific role of each component can be found in the corresponding technical report of the European Joint Research Center (JRC).

We also offer a MOSH/MOAH retention time CRM mix to accurately cut and quantify subfractions of MOSH and MOAH.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Perylene
    5-α-Cholestane
    n-Undecane
    n-Tridecane
    Tri-tert-butylbenzene
    Bicyclohexyl
    1-Methylnaphthalene
    2-Methylnaphthalene
    Pentylbenzene
    1,4-Bis-(2-ethylhexyl)benzene
    See All (10)

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.9 °F

Flash Point(C)

4.4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service