Skip to Content
Merck
All Photos(1)

Key Documents

SBP3957

Sigma-Aldrich

Slug (AB1) Blocking Peptide for PRS3957

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

synthetic

form

liquid

species reactivity

mouse, human

concentration

200 μg/mL

shipped in

wet ice

storage temp.

−20°C

Immunogen

14 amino acids near the center of human Slug.

Application

Used as a blocking peptide in immunoblotting applications.

Linkage

This peptide blocks the action of antibody PRS3957.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service