SAB5202683
Monoclonal Anti-Sulfotyrosine-Horseradish Peroxidase antibody produced in mouse
clone 7C5
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.45
Recommended Products
biological source
mouse
conjugate
peroxidase conjugate
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
7C5, monoclonal
form
buffered aqueous solution
species reactivity
human
concentration
1.0 mg/mL
technique(s)
immunofluorescence: 1:50
western blot: 1:1000
isotype
IgG1
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
sulfation (Tyr)
Immunogen
Synthetic Sulfotyrosine conjugated to Keyhole Limpet Hemocyanin (KLH).
Physical form
PBS pH 7.4, 50% glyercol, 0.09% Sodium Azide
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service