Skip to Content
Merck
All Photos(1)

Key Documents

PZ0112

Sigma-Aldrich

PD-184161

≥98% (HPLC)

Synonym(s):

5-Bromo-2-[(2-chloro-4-iodophenyl)amino]-N-(cyclopropylmethoxy)-3,4-difluoro-benzamide, PF-1529483, PF-3011370, UK-287074

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H13BrClF2IN2O2
CAS Number:
Molecular Weight:
557.56
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

Fc1c(F)c(Nc2ccc(I)cc2Cl)c(cc1Br)C(=O)NOCC3CC3

InChI

1S/C17H13BrClF2IN2O2/c18-11-6-10(17(25)24-26-7-8-1-2-8)16(15(21)14(11)20)23-13-4-3-9(22)5-12(13)19/h3-6,8,23H,1-2,7H2,(H,24,25)

InChI key

VJNZMSLGVUSPCF-UHFFFAOYSA-N

Biochem/physiol Actions

PD-184161 is a MEK inhibitor.

Features and Benefits

This compound is featured on the MAPKKKs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akikazu Harada et al.
eLife, 10 (2021-10-01)
Pancreatic cancer has a high mortality rate due to metastasis. Whereas KRAS is mutated in most pancreatic cancer patients, controlling KRAS or its downstream effectors has not been succeeded clinically. ARL4C is a small G protein whose expression is induced
A Dushani C U Ranasinghe et al.
Biochimica et biophysica acta. Molecular and cell biology of lipids, 1865(12), 158789-158789 (2020-08-11)
Phosphorylation of sphingosine by sphingosine kinase 1 (SPHK1) produces the bioactive sphingolipid sphingosine-1-phosphate (S1P), a microvascular and immuno-modulator associated with vascular remodeling in pulmonary arterial hypertension (PAH). The low intracellular concentration of S1P is under tight spatial-temporal control. Molecular mechanisms
Zhengxi Wei et al.
Toxicology and applied pharmacology, 289(1), 98-108 (2015-09-20)
Cadmium (Cd) is a carcinogenic metal which is implicated in breast cancer by epidemiological studies. It is reported to promote breast cancer cell growth in vitro through membrane receptors. The study described here examined Cd-mediated growth of non-metastatic human breast

Articles

The extracellular signal regulated kinase (ERK1 and ERK2) pathways are activated by mitogens and play an important role in controlling cell growth and differentiation.

The extracellular signal regulated kinase (ERK1 and ERK2) pathways are activated by mitogens and play an important role in controlling cell growth and differentiation.

The extracellular signal regulated kinase (ERK1 and ERK2) pathways are activated by mitogens and play an important role in controlling cell growth and differentiation.

The extracellular signal regulated kinase (ERK1 and ERK2) pathways are activated by mitogens and play an important role in controlling cell growth and differentiation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service