Key Documents
MTCP4408
MTC Bio SureStand Multiple Pipette Stand
to hold, 8 x single pipettes OR 6 x multipipettes
Synonym(s):
pipette holder, pipette rack, plastic tube
About This Item
UNSPSC Code:
41122801
NACRES:
NB.02
material
acrylic
feature
to hold 8 x single pipettes OR 6 x multipipettes
packaging
pack of 1 ea
manufacturer/tradename
MTC Bio P4408
General description
SureStand Pipette Stand for 8 pipettes, up to six multi-channels, acrylic
SureStand Pipette racks are unique in their ability to accommodate multi-channel pipettes without sacrificing any of the stability of a traditional acrylic rack. The patented “windows” allow the user to insert the multi-channel head longways through the rack while keeping the body of the pipette stable by securing the pipette at two points: under the finger hook and near the bottom of the handle. Each SureStand is also outfitted with four non-skid rubber “feet” for extra stability. SureStand is guaranteed to be the sturdiest and most versatile pipette rack in your lab.
Features and Benefits
  • Universal – Fits all popular pipettes
  • 5mm acrylic – 40% thicker
  • Non-skid rubber feet
  • Multi-channel “window” slots
  • US Patent No 872,300
Legal Information
MTC is a trademark of MTC Bio
SureStand is a trademark of MTC Bio
Choose from one of the most recent versions:
Certificates of Analysis (COA)