Skip to Content
Merck
All Photos(1)

Key Documents

EHU121641

Sigma-Aldrich

MISSION® esiRNA

targeting human FZD7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCTGGCCTGCTACTTCTACGAGCAGGCCTTCCGCGAGCACTGGGAGCGCACCTGGCTCCTGCAGACGTGCAAGAGCTATGCCGTGCCCTGCCCGCCCGGCCACTTCCCGCCCATGAGCCCCGACTTCACCGTCTTCATGATCAAGTACCTGATGACCATGATCGTCGGCATCACCACTGGCTTCTGGATCTGGTCGGGCAAGACCCTGCAGTCGTGGCGCCGCTTCTACCACAGACTTAGCCACAGCAGCAAGGGGGAGACTGCGGTATGAGCCCCGGCCCCTCCCCACCTTTCCCACCCCAGCCCTCTTGCAAGAGGAGAGGCACGGTAGGGAAAAGAACTGCTGGGTGGGGGCCTGTTTCTGTAACTTTCTCCCCCTCTACTGAGAAGTGACCTGGAAGTGAGAAGTTCTTTGCAGATTTGGGGCGAGGGGTGATTTGGAAAAGAAGACCTGGGTG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Geng et al.
Oncology reports, 35(4), 2441-2450 (2016-01-20)
MicroRNAs (miRNAs) are novel tools for cancer therapy. Frizzled7 (FZD7) is an important co-receptor in the WNT signaling pathway. The WNT signaling pathway is aberrantly activated in Helicobacter pylori (H. pylori)‑infected gastric cancer cells. However, the role of FZD7 in
Yufeng Wang et al.
Cell death & disease, 9(9), 851-851 (2018-08-30)
Previous evidences reveal that long non-coding RNA (lncRNA) down syndrome critical region 8 (DSCR8) involves in the progression of multiple cancers. However, the exact expression, function, and mechanism of DSCR8 in hepatocellular carcinoma (HCC) remain uncovered. In this study, real-time
M Asad et al.
Cell death & disease, 5, e1346-e1346 (2014-07-18)
Ovarian cancer (OC) can be classified into five biologically distinct molecular subgroups: epithelial-A (Epi-A), Epi-B, mesenchymal (Mes), Stem-A and Stem-B. Among them, Stem-A expresses genes relating to stemness and is correlated with poor clinical prognosis. In this study, we show
Salvatore Condello et al.
Cancer research, 78(11), 2990-3001 (2018-03-08)
Cancer progression and recurrence are linked to a rare population of cancer stem cells (CSC). Here, we hypothesized that interactions with the extracellular matrix drive CSC proliferation and tumor-initiating capacity and investigated the functions of scaffold protein tissue transglutaminase (TG2)
Mio Nakanishi et al.
Cell, 177(4), 910-924 (2019-04-16)
The assembly of organized colonies is the earliest manifestation in the derivation or induction of pluripotency in vitro. However, the necessity and origin of this assemblance is unknown. Here, we identify human pluripotent founder cells (hPFCs) that initiate, as well as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service