Skip to Content
Merck
All Photos(1)

Key Documents

Eos-HL-60

96100920, human blood, Lymphoblastoid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

Eos-HL-60, 96100920

biological source

human blood

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblastoid

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian promyelocytic leukaemia

Cell Line Description

Eos-HL-60 is a variant of the promyelocytic cell line HL60 established by sub-cloning in methylcellulose. Although the cells are capable of reverting to the parental phenotype, most passages maintain a high degree of eosinophil differentiation.Eos-HL-60 and HL-60 have been shown to be identical genetically by STR profiling

Application

Study of production and function of human eosinophils, analysis of differentiation.

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 13,14
D13S317: 8,11
D16S539: 11
D5S818: 12
D7S820: 11,12
THO1: 8
TPOX: 8,11
vWA: 16,17

Culture Medium

IDMEM + 2mM Glutamine + 0.5mM Butyric acid + 10% Foetal Bovine Serum (FBS); pH7.6; add Butyric acid to the medium just before use.

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service