Skip to Content
Merck
All Photos(1)

Documents

LS174T

87060401, human colon, Epithelial

Synonym(s):

LS-174-T Cells, LS-174T Cells, LS174 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

LS174T, 87060401

biological source

human colon

growth mode

Adherent

karyotype

Modal no. 45 (2n

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian colon adenocarcinoma

Cell Line Description

A trypsinised variant of LS180 (Sigma Catalogue number. 87021202), both lines being derived from a 58 year old female with Dukes type B adenocarcinoma of the colon. The cells have microvilli and intracytoplasmic vacuoles. The cells are tumorigenic in nude mice.

Application

LS174T is part of the three-dimensional (3D) heterotypic spheroid model developed to assess cancer immunotherapy agents in vitro. It has been used to study the interaction of enterohaemorrhagic Escherichia coli (EHEC) with the intestinal mucus layer produced by LS174T cells.

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:5 i.e. seeding at 2-5x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C May take up to 7 days for cells to attach. Cells grow in islands and tend to pile on top of each other. Some cells ma

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service