Skip to Content
Merck
All Photos(1)

Key Documents

CGR8

7032901, mouse embryo, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
mouse embryo
growth mode:
Adherent
karyotype:
40XY
morphology:
Not specified
products:
Not specified
receptors:
Not specified

Product Name

CGR8, 07032901

biological source

mouse embryo

growth mode

Adherent

karyotype

40XY

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse embryonic stem cell

Cell Line Description

The germ-line competent cell line CGR8 was established from the inner cell mass of a 3.5 day male pre-implantation mouse embryo (Mus musculus, strain 129). These pluripotent cells retain the ability to participate in normal embryonic development. Differentiation of CGR8 cells is inhibited by the pleiotropic cytokine Differentiation Inhibiting Activity (DIA) which is identical to Leukaemia Inhibiting Factor (LIF). Addition of DIA/LIF allows culture of CGR8 without the use of feeder layers. Cells are small and tightly packed.

Application

Gene targeting, Gene trapping, in vitro differentiation

Culture Medium

GMEM + 2mM Glutamine + 0.05mM 2-Mercaptoethanol (2ME) + 1000 units/ml DIA/LIF + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:10 i.e. seeding at 4x1000 cells/cm² using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. CGR8 cells should be cultured on gelatin - coated flasks. Flasks should be coated using 0.2% gelatin in PBS. ECACC introduced the use of a feeder layer of mitomycin C treated primary mouse embryonic fibroblast (PMEF) cells for the bulk culture of CGR8. This means the ampoules that we provide contain the PMEF feeder cells as well as the CGR8 cells. When the cells are initially resuscitated and plated out from the ampoule both the CGR8 stem cell colonies and the fibroblast feeder cells will be visible in the culture. Feeder layers are not essential and the use of LIF at the correct concentration (with daily media changes) should be sufficient to maintain the pluripotency of the cells in end user laboratories.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service