Skip to Content
Merck
All Photos(1)

Key Documents

Y0001125

Orbifloxacin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-Cyclopropyl-5,6,8-trifluoro-1,4-dihydro-7-(cis-3,5-dimethyl-1-piperazinyl)-4-oxo-quinoline-3-carboxylic acid, Orbax

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20F3N3O3
CAS Number:
Molecular Weight:
395.38
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

orbifloxacin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

C[C@@H]1CN(C[C@H](C)N1)c2c(F)c(F)c3C(=O)C(=CN(C4CC4)c3c2F)C(O)=O

InChI

1S/C19H20F3N3O3/c1-8-5-24(6-9(2)23-8)17-14(21)13(20)12-16(15(17)22)25(10-3-4-10)7-11(18(12)26)19(27)28/h7-10,23H,3-6H2,1-2H3,(H,27,28)/t8-,9+

InChI key

QIPQASLPWJVQMH-DTORHVGOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Orbifloxacin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tao Wang et al.
Arteriosclerosis, thrombosis, and vascular biology, 35(5), 1218-1225 (2015-04-04)
Surgical hindlimb ischemia (HLI) in mice has become a valuable preclinical model to study peripheral arterial disease. We previously identified that the different phenotypic outcomes after HLI across inbred mouse strains is related to a region on the short arm
Sarah M Churchman et al.
Arthritis research & therapy, 16(6), 511-511 (2014-12-24)
Interleukin-7 (IL-7) is a cytokine essential for T-cell lymphopoiesis, survival and polarization with an emerging role in autoimmunity. We previously demonstrated reduced levels of circulating IL-7 in rheumatoid arthritis (RA), although high amounts are expressed in joints, suggesting differences between
Lindsey L Kennedy et al.
Laboratory investigation; a journal of technical methods and pathology, 94(12), 1406-1418 (2014-11-05)
Cholangiopathies are characterized by dysregulation of the balance between biliary growth and loss. We have shown that histamine (HA) stimulates biliary growth via autocrine mechanisms. To evaluate the paracrine effects of mast cell (MC) stabilization on biliary proliferation, sham or
Xiaoqiang Liu et al.
Veterinary microbiology, 174(3-4), 514-522 (2014-12-04)
This study explored and compared the mechanisms and selective concentration of resistance between a 3rd (pradofloxacin) and 2nd (ciprofloxacin) generation fluoroquinolone. Pradofloxacin- and ciprofloxacin-resistant mutants were selected by stepwise exposure of Escherichia coli (E. coli) to escalating concentrations of pradofloxacin
Abdelouahid El-Khattouti et al.
Cancer letters, 357(1), 83-104 (2014-12-03)
According to the cancer stem-like cell (CSC) hypothesis, neoplastic clones are maintained by a small fraction of cells with stem cell properties. Also, melanoma resistance to chemo- and radiotherapy is thought to be attributed to melanoma stem-like cells (MSCs). Caffeic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service