PHR8709
Fosinopril Related Compound A
certified reference material, pharmaceutical secondary standard
Synonym(s):
Fosinoprilat, (4S)-4-Cyclohexyl-1-{[hydroxy(4-phenylbutyl)phosphinyl]acetyl}-L-proline
About This Item
Recommended Products
grade
certified reference material
pharmaceutical secondary standard
CofA
current certificate can be downloaded
packaging
pkg of 50 mg
application(s)
pharmaceutical
SMILES string
[P](=O)(O)(CCCCc3ccccc3)CC(=O)N1[C@@H](C[C@H](C1)C2CCCCC2)C(=O)O
InChI
1S/C23H34NO5P/c25-22(17-30(28,29)14-8-7-11-18-9-3-1-4-10-18)24-16-20(15-21(24)23(26)27)19-12-5-2-6-13-19/h1,3-4,9-10,19-21H,2,5-8,11-17H2,(H,26,27)(H,28,29)/t20-,21+/m1/s1
InChI key
WOIWWYDXDVSWAZ-RTWAWAEBSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Analysis Note
Other Notes
Footnote
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Repr. 1A
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service